Assay Details
| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078046.1 | NP_001071514.1 | 11 - 12 | 1298 | 77 | |
| GenBank mRNA | BC126607.1 | - | 11 - 12 | 1298 | 77 | |
More Information
Set Membership: |
|

CDS2
Genomic Map