Assay Details
| Gene Symbol: GSDMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078090.1 | NP_001071558.1 | 7 - 8 | 806 | 61 | |
| GenBank mRNA | BC126753.1 | - | 7 - 8 | 806 | 61 | |
More Information
Set Membership: |
|

GSDMA
Genomic Map