Assay Details
| Gene Symbol: MPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079507.2 | NP_001072975.1 | 1 - 2 | 121 | 76 | |
| NM_001316359.1 | NP_001303288.1 | 1 - 2 | 121 | 76 | ||
| GenBank mRNA | BC122659.1 | - | 1 - 2 | 113 | 76 | |
| EE369960.1 | - | 1 - 2 | 121 | 76 | ||
More Information
Set Membership: |
|

MPZ
Genomic Map