Assay Details
| Gene Symbol: TREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079580.2 | NP_001073048.1 | 1 - 2 | 72 | 77 | |
| GenBank mRNA | BC122771.1 | - | 1 - 2 | 73 | 77 | |
More Information
Set Membership: |
|

TREM2
Genomic Map