Assay Details
| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079609.1 | NP_001073077.1 | 7 - 8 | 1128 | 83 | |
| GenBank mRNA | BC126507.1 | - | 7 - 8 | 1128 | 83 | |
More Information
Set Membership: |
|

CA10
Genomic Map