Assay Details
| Gene Symbol: METTL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079782.2 | NP_001073250.1 | 4 - 5 | 712 | 57 | |
| GenBank mRNA | BC126575.1 | - | 4 - 5 | 713 | 57 | |
More Information
Set Membership: |
|

METTL15
Genomic Map