Assay Details
| Gene Symbol: TFAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080228.1 | NP_001073697.1 | 2 - 3 | 518 | 70 | |
| GenBank mRNA | BC126845.1 | - | 2 - 3 | 518 | 70 | |
More Information
Set Membership: |
|

TFAP2A
Genomic Map