Assay Details
| Gene Symbol: THG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080233.2 | NP_001073702.1 | 1 - 2 | 207 | 86 | |
| GenBank mRNA | BC122826.1 | - | 1 - 2 | 210 | 86 | |
More Information
Set Membership: |
|

THG1L
Genomic Map