Assay Details
| Gene Symbol: ZCCHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080234.1 | NP_001073703.1 | 1 | 281 | 139 | |
| GenBank mRNA | BC126595.1 | - | 1 | 281 | 139 | |
More Information
Set Membership: |
|

ZCCHC3
Genomic Map