Assay Details
| Gene Symbol: SCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080243.1 | NP_001073712.1 | 1 - 2 | 312 | 57 | |
| GenBank mRNA | BC126613.1 | - | 1 - 2 | 312 | 57 | |
More Information
Set Membership: |
|

SCO1
Genomic Map