Assay Details
| Gene Symbol: TIMM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080277.1 | NP_001073746.1 | 2 - 3 | 201 | 101 | |
| GenBank mRNA | BC126713.1 | - | 2 - 3 | 201 | 101 | |
More Information
Set Membership: |
|

TIMM13
Genomic Map