Assay Details
| Gene Symbol: SLC20A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080280.1 | NP_001073749.1 | 3 - 4 | 954 | 66 | |
| GenBank mRNA | BC126506.1 | - | 3 - 4 | 954 | 66 | |
More Information
Set Membership: |
|

SLC20A2
Genomic Map