Assay Details
| Gene Symbol: MEPCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080283.2 | NP_001073752.2 | 1 - 2 | 1759 | 65 | |
| GenBank mRNA | BT029808.1 | - | 1 - 2 | 988 | 65 | |
| EE245239.1 | - | 1 - 2 | 371 | 65 | ||
More Information
Set Membership: |
|

MEPCE
Genomic Map