Assay Details
| Gene Symbol: EFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080299.1 | NP_001073768.1 | 4 - 5 | 1379 | 66 | |
| GenBank mRNA | BT029857.1 | - | 4 - 5 | 1379 | 66 | |
More Information
Set Membership: |
|

EFNB1
Genomic Map