Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080319.2 | NP_001073788.1 | 16 - 17 | 2205 | 75 | |
| GenBank mRNA | BC126671.1 | - | 16 - 17 | 2206 | 75 | |
More Information
Set Membership: |
|

MINDY4
Genomic Map