Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080319.2 | NP_001073788.1 | 2 - 3 | 220 | 149 | |
| GenBank mRNA | BC126671.1 | - | 2 - 3 | 221 | 149 | |
More Information
Set Membership: |
|

MINDY4
Genomic Map