Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080319.2 | NP_001073788.1 | 7 - 8 | 1295 | 73 | |
| GenBank mRNA | BC126671.1 | - | 7 - 8 | 1296 | 73 | |
More Information
Set Membership: |
|

MINDY4
Genomic Map