Assay Details
| Gene Symbol: OVOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081521.1 | NP_001074990.1 | 2 - 3 | 586 | 74 | |
| GenBank mRNA | BC133390.1 | - | 2 - 3 | 586 | 74 | |
More Information
Set Membership: |
|

OVOL1
Genomic Map