Assay Details
Gene Symbol: HABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081523.1 | NP_001074992.1 | 7 - 8 | 1292 | 70 | |
GenBank mRNA | BC133323.1 | - | 7 - 8 | 1292 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
