Assay Details
| Gene Symbol: NXPE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081529.2 | NP_001074998.1 | 5 - 6 | 1179 | 73 | |
| GenBank mRNA | BC133340.1 | - | 5 - 6 | 1180 | 73 | |
More Information
Set Membership: |
|

NXPE3
Genomic Map