Assay Details
| Gene Symbol: ENPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081535.1 | NP_001075004.1 | 3 - 4 | 1057 | 97 | |
| GenBank mRNA | BC133338.1 | - | 3 - 4 | 1057 | 97 | |
More Information
Set Membership: |
|

ENPP4
Genomic Map