Assay Details
| Gene Symbol: ITM2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081538.1 | NP_001075007.1 | 3 - 4 | 524 | 67 | |
| GenBank mRNA | BC133321.1 | - | 3 - 4 | 524 | 67 | |
More Information
Set Membership: |
|

ITM2C
Genomic Map