Assay Details
| Gene Symbol: ELK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081542.1 | NP_001075011.1 | 4 - 5 | 1316 | 90 | |
| GenBank mRNA | BC133391.1 | - | 4 - 5 | 1316 | 90 | |
More Information
Set Membership: |
|

ELK3
Genomic Map