Assay Details
| Gene Symbol: RAB40C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081545.1 | NP_001075014.1 | 3 - 4 | 374 | 67 | |
| GenBank mRNA | BC133361.1 | - | 3 - 4 | 374 | 67 | |
More Information
Set Membership: |
|

RAB40C
Genomic Map