Assay Details
| Gene Symbol: ZNF567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081599.1 | NP_001075068.1 | 3 - 4 | 302 | 113 | |
| GenBank mRNA | BC133337.1 | - | 3 - 4 | 302 | 113 | |
More Information
Set Membership: |
|

ZNF567
Genomic Map