Assay Details
| Gene Symbol: TMED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081609.2 | NP_001075078.1 | 2 - 3 | 556 | 130 | |
| GenBank mRNA | BC133434.1 | - | 2 - 3 | 557 | 130 | |
More Information
Set Membership: |
|

TMED3
Genomic Map