Assay Details
| Gene Symbol: ADAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081620.1 | NP_001075089.1 | 4 - 5 | 487 | 66 | |
| GenBank mRNA | BC133289.1 | - | 4 - 5 | 487 | 66 | |
More Information
Set Membership: |
|

ADAT1
Genomic Map