Assay Details
| Gene Symbol: SAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081711.1 | NP_001075180.1 | 4 - 5 | 599 | 64 | |
| GenBank mRNA | BC133519.1 | - | 4 - 5 | 599 | 64 | |
More Information
Set Membership: |
|

SAE1
Genomic Map