Assay Details
| Gene Symbol: SERPINA3-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081712.1 | NP_001075181.1 | 4 - 5 | 1098 | 54 | |
| GenBank mRNA | BC149427.1 | - | 4 - 5 | 1148 | 54 | |
More Information
Set Membership: |
|

SERPINA3-8
Genomic Map