Assay Details
| Gene Symbol: CLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081713.1 | NP_001075182.1 | 2 - 3 | 679 | 90 | |
| GenBank mRNA | BC133499.1 | - | 2 - 3 | 679 | 90 | |
More Information
Set Membership: |
|

CLP1
Genomic Map