Assay Details
| Gene Symbol: FAM8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081718.1 | NP_001075187.1 | 3 - 4 | 1043 | 121 | |
| GenBank mRNA | BC133604.1 | - | 3 - 4 | 1043 | 121 | |
More Information
Set Membership: |
|

FAM8A1
Genomic Map