Assay Details
| Gene Symbol: MAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081719.1 | NP_001075188.1 | 2 - 3 | 382 | 82 | |
| GenBank mRNA | BC133516.1 | - | 2 - 3 | 382 | 82 | |
More Information
Set Membership: |
|

MAL2
Genomic Map