Assay Details
| Gene Symbol: CLIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081727.1 | NP_001075196.1 | 1 - 2 | 307 | 78 | |
| GenBank mRNA | BC133567.1 | - | 1 - 2 | 307 | 78 | |
More Information
Set Membership: |
|

CLIC2
Genomic Map