Assay Details
| Gene Symbol: PPP2R5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081728.1 | NP_001075197.1 | 1 - 2 | 695 | 73 | |
| GenBank mRNA | BC133482.1 | - | 1 - 2 | 695 | 73 | |
More Information
Set Membership: |
|

PPP2R5A
Genomic Map