Assay Details
| Gene Symbol: IKBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081736.2 | NP_001075205.1 | 1 - 2 | 253 | 149 | |
| GenBank mRNA | BC133577.1 | - | 1 - 2 | 254 | 149 | |
More Information
Set Membership: |
|

IKBIP
Genomic Map