Assay Details
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082425.1 | NP_001075894.1 | 1 - 2 | 176 | 109 | |
| GenBank mRNA | BC133571.1 | - | 1 - 2 | 176 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC133571.1, NM_001082425.1 transcripts.
Set Membership: |
|

KLF15
Genomic Map