Assay Details
| Gene Symbol: STX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082431.1 | NP_001075900.1 | 4 - 5 | 553 | 62 | |
| GenBank mRNA | BC133525.1 | - | 4 - 5 | 553 | 62 | |
More Information
Set Membership: |
|

STX6
Genomic Map