Assay Details
| Gene Symbol: COX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082437.2 | NP_001075906.1 | 2 - 3 | 505 | 77 | |
| GenBank mRNA | BC133634.1 | - | 2 - 3 | 506 | 77 | |
More Information
Set Membership: |
|

COX18
Genomic Map