Assay Details
| Gene Symbol: FBXL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082440.1 | NP_001075909.1 | 8 - 9 | 1168 | 73 | |
| GenBank mRNA | BC133613.1 | - | 8 - 9 | 1168 | 73 | |
More Information
Set Membership: |
|

FBXL5
Genomic Map