Assay Details
| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082447.2 | NP_001075916.1 | 6 - 7 | 939 | 72 | |
| GenBank mRNA | BC133640.1 | - | 6 - 7 | 940 | 72 | |
More Information
Set Membership: |
|

SEPTIN11
Genomic Map