Assay Details
| Gene Symbol: RHOBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082453.2 | NP_001075922.1 | 2 - 3 | 357 | 60 | |
| GenBank mRNA | BC133510.1 | - | 2 - 3 | 358 | 60 | |
More Information
Set Membership: |
|

RHOBTB1
Genomic Map