Assay Details
| Gene Symbol: NOCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082454.2 | NP_001075923.1 | 2 - 3 | 676 | 71 | |
| GenBank mRNA | BC133608.1 | - | 2 - 3 | 677 | 71 | |
| BC149515.1 | - | 4 - 5 | 512 | 71 | ||
More Information
Set Membership: |
|

NOCT
Genomic Map