Assay Details
Gene Symbol: UNC93A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082457.1 | NP_001075926.1 | 7 - 8 | 1266 | 64 | |
GenBank mRNA | BC133579.1 | - | 7 - 8 | 1266 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
