Assay Details
| Gene Symbol: SAPCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082467.1 | NP_001075936.1 | 3 - 4 | 604 | 79 | |
| GenBank mRNA | BC133521.1 | - | 3 - 4 | 604 | 79 | |
More Information
Set Membership: |
|

SAPCD1
Genomic Map