Assay Details
| Gene Symbol: GPR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082470.2 | NP_001075939.1 | 1 - 2 | 61 | 80 | |
| GenBank mRNA | BC133644.1 | - | 1 - 2 | 62 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC133644.1, NM_001082470.2 transcripts.
Set Membership: |
|

GPR17
Genomic Map