Assay Details
| Gene Symbol: EXTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082473.1 | NP_001075942.1 | 4 - 5 | 2841 | 64 | |
| GenBank mRNA | BC133540.1 | - | 4 - 5 | 2841 | 64 | |
More Information
Set Membership: |
|

EXTL3
Genomic Map