Assay Details
| Gene Symbol: DUSP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082609.2 | NP_001076078.1 | 2 - 3 | 292 | 74 | |
| GenBank mRNA | BC133448.1 | - | 2 - 3 | 293 | 74 | |
| BC134731.1 | - | 2 - 3 | 303 | 74 | ||
More Information
Set Membership: |
|

DUSP23
Genomic Map