Assay Details
| Gene Symbol: DKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082615.1 | NP_001076084.1 | 1 - 2 | 923 | 80 | |
| GenBank mRNA | BC133394.1 | - | 1 - 2 | 923 | 80 | |
More Information
Set Membership: |
|

DKK2
Genomic Map