Assay Details
| Gene Symbol: KRT82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082616.1 | NP_001076085.1 | 5 - 6 | 954 | 54 | |
| GenBank mRNA | BC133393.1 | - | 5 - 6 | 954 | 54 | |
More Information
Set Membership: |
|

KRT82
Genomic Map