Assay Details
| Gene Symbol: TRIR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083371.2 | NP_001076840.1 | 2 - 3 | 454 | 66 | |
| GenBank mRNA | BC133464.1 | - | 2 - 3 | 455 | 66 | |
More Information
Set Membership: |
|

TRIR
Genomic Map